Assay Details
| Gene Symbol: SBDS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034439.1 | NP_001029611.1 | 1 - 2 | 218 | 78 | |
| GenBank mRNA | BC104585.1 | - | 1 - 2 | 218 | 78 | |
More Information
Set Membership: |
|

SBDS
Genomic Map