Assay Details
| Gene Symbol: CCDC115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034444.1 | NP_001029616.1 | 3 - 4 | 294 | 133 | |
| GenBank mRNA | BC102977.1 | - | 3 - 4 | 294 | 133 | |
More Information
Set Membership: |
|

CCDC115
Genomic Map