Assay Details
| Gene Symbol: KPNA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034449.2 | NP_001029621.1 | 3 - 4 | 335 | 100 | |
| GenBank mRNA | BC103363.1 | - | 3 - 4 | 336 | 100 | |
More Information
Set Membership: |
|

KPNA2
Genomic Map