Assay Details
| Gene Symbol: VPS29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034455.2 | NP_001029627.1 | 3 - 4 | 248 | 72 | |
| GenBank mRNA | BC102341.1 | - | 3 - 4 | 250 | 72 | |
More Information
Set Membership: |
|

VPS29
Genomic Map