Assay Details
| Gene Symbol: QARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034468.2 | NP_001029640.1 | 20 - 21 | 1976 | 72 | |
| GenBank mRNA | BC105237.1 | - | 20 - 21 | 1977 | 72 | |
| BT025487.1 | - | 20 - 21 | 1963 | 72 | ||
More Information
Set Membership: |
|

QARS1
Genomic Map