Assay Details
| Gene Symbol: MESD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034469.1 | NP_001029641.1 | 2 - 3 | 460 | 68 | |
| GenBank mRNA | BC102262.1 | - | 2 - 3 | 460 | 68 | |
More Information
Set Membership: |
|

MESD
Genomic Map