Assay Details
| Gene Symbol: GDPD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034471.2 | NP_001029643.2 | 10 - 11 | 993 | 82 | |
| GenBank mRNA | BC102804.1 | - | 7 - 8 | 1203 | 82 | |
More Information
Set Membership: |
|

GDPD2
Genomic Map