Assay Details
| Gene Symbol: CDIPT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034482.1 | NP_001029654.1 | 1 - 2 | 390 | 91 | |
| GenBank mRNA | BC102187.1 | - | 1 - 2 | 390 | 91 | |
More Information
Set Membership: |
|

CDIPT
Genomic Map