Assay Details
| Gene Symbol: C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034492.1 | NP_001029664.1 | 8 - 9 | 1166 | 58 | |
| GenBank mRNA | BC103357.1 | - | 8 - 9 | 1166 | 58 | |
| BT026538.1 | - | 8 - 9 | 1149 | 58 | ||
More Information
Set Membership: |
|

C2
Genomic Map