Assay Details
| Gene Symbol: RABL2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034508.2 | NP_001029680.1 | 1 - 2 | 169 | 76 | |
| GenBank mRNA | BC102718.1 | - | 1 - 2 | 170 | 76 | |
More Information
Set Membership: |
|

RABL2B
Genomic Map