Assay Details
| Gene Symbol: NUDT16L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034526.1 | NP_001029698.1 | 5 - 6 | 1044 | 59 | |
| GenBank mRNA | BC104543.1 | - | 5 - 6 | 1044 | 59 | |
More Information
Set Membership: |
|

NUDT16L1
Genomic Map