Assay Details
| Gene Symbol: NSA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034546.2 | NP_001029718.1 | 2 - 3 | 303 | 121 | |
| GenBank mRNA | BC104563.1 | - | 2 - 3 | 304 | 121 | |
More Information
Set Membership: |
|

NSA2
Genomic Map