Assay Details
| Gene Symbol: SLC31A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034556.1 | NP_001029728.1 | 3 - 4 | 375 | 67 | |
| GenBank mRNA | BC102362.1 | - | 3 - 4 | 375 | 67 | |
More Information
Set Membership: |
|

SLC31A2
Genomic Map