Assay Details
| Gene Symbol: CIDEB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034567.2 | NP_001029739.1 | 1 - 2 | 127 | 72 | |
| GenBank mRNA | BC102065.1 | - | 1 - 2 | 128 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102065.1, NM_001034567.2 transcripts.
Set Membership: |
|

CIDEB
Genomic Map