Assay Details
| Gene Symbol: CIDEB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034567.2 | NP_001029739.1 | 4 - 5 | 612 | 103 | |
| GenBank mRNA | BC102065.1 | - | 4 - 5 | 613 | 103 | |
More Information
Set Membership: |
|

CIDEB
Genomic Map