Assay Details
| Gene Symbol: C1GALT1C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034575.1 | NP_001029747.1 | 1 - 2 | 167 | 93 | |
| GenBank mRNA | BC104496.1 | - | 1 - 2 | 167 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC104496.1, NM_001034575.1 transcripts.
Set Membership: |
|

C1GALT1C1
Genomic Map