Assay Details
| Gene Symbol: PSMB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034596.2 | NP_001029768.1 | 3 - 4 | 383 | 60 | |
| GenBank mRNA | BC102554.1 | - | 3 - 4 | 384 | 60 | |
| BC151435.1 | - | 3 - 4 | 383 | 60 | ||
More Information
Set Membership: |
|

PSMB3
Genomic Map