Assay Details
| Gene Symbol: PDZK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034611.2 | NP_001029783.1 | 1295 | 63 | ||
| GenBank mRNA | BC102213.1 | - | 1296 | 63 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PDZK1| Gene Symbol: PDZK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034611.2 | NP_001029783.1 | 1295 | 63 | ||
| GenBank mRNA | BC102213.1 | - | 1296 | 63 | ||
Set Membership: |
|