Assay Details
| Gene Symbol: PMPCB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034613.1 | NP_001029785.1 | 8 - 9 | 995 | 57 | |
| GenBank mRNA | BC103085.1 | - | 8 - 9 | 995 | 57 | |
More Information
Set Membership: |
|

PMPCB
Genomic Map