Assay Details
| Gene Symbol: EIF3CL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034618.2 | NP_001029790.1 | 1 - 2 | 53 | 110 | |
| GenBank mRNA | BC103352.1 | - | 1 - 2 | 54 | 110 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC103352.1, NM_001034618.2 transcripts.
Set Membership: |
|

EIF3CL
Genomic Map