Assay Details
| Gene Symbol: ANKRD46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034619.1 | NP_001029791.1 | 3 - 4 | 526 | 102 | |
| GenBank mRNA | BC104580.1 | - | 3 - 4 | 526 | 102 | |
More Information
Set Membership: |
|

ANKRD46
Genomic Map