Assay Details
| Gene Symbol: SYNGR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034623.1 | NP_001029795.1 | 2 - 3 | 366 | 77 | |
| GenBank mRNA | BC103195.1 | - | 2 - 3 | 366 | 77 | |
More Information
Set Membership: |
|

SYNGR1
Genomic Map