Assay Details
| Gene Symbol: RPL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034625.2 | NP_001029797.1 | 1 | 310 | 55 | |
| GenBank mRNA | BC102277.1 | - | 1 | 311 | 55 | |
| BC151407.1 | - | 3 - 4 | 312 | 55 | ||
More Information
Set Membership: |
|

RPL8
Genomic Map