Assay Details
| Gene Symbol: RPL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034625.2 | NP_001029797.1 | 1 | 654 | 127 | |
| GenBank mRNA | BC102277.1 | - | 1 | 655 | 127 | |
| BC151407.1 | - | 5 - 6 | 656 | 127 | ||
More Information
Set Membership: |
|

RPL8
Genomic Map