Assay Details
| Gene Symbol: GRB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034630.1 | NP_001029802.1 | 5 - 6 | 805 | 66 | |
| GenBank mRNA | BC102411.1 | - | 5 - 6 | 805 | 66 | |
More Information
Set Membership: |
|

GRB2
Genomic Map