Assay Details
| Gene Symbol: RAB28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034634.1 | NP_001029806.1 | 1 - 2 | 288 | 88 | |
| GenBank mRNA | BC104592.1 | - | 1 - 2 | 288 | 88 | |
More Information
Set Membership: |
|

RAB28
Genomic Map