Assay Details
| Gene Symbol: SLC35A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034637.2 | NP_001029809.1 | 7 - 8 | 915 | 108 | |
| GenBank mRNA | BC102765.1 | - | 7 - 8 | 916 | 108 | |
More Information
Set Membership: |
|

SLC35A1
Genomic Map