Assay Details
| Gene Symbol: SUCLG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034639.1 | NP_001029811.1 | 5 - 6 | 571 | 71 | |
| GenBank mRNA | BC104559.1 | - | 5 - 6 | 590 | 71 | |
| BT030582.1 | - | 5 - 6 | 568 | 71 | ||
More Information
Set Membership: |
|

SUCLG2
Genomic Map