Assay Details
| Gene Symbol: CDH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034640.2 | NP_001029812.1 | 3 - 4 | 737 | 60 | |
| GenBank mRNA | BC104618.1 | - | 3 - 4 | 739 | 60 | |
More Information
Set Membership: |
|

CDH6
Genomic Map