Assay Details
| Gene Symbol: MRPL16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034641.2 | NP_001029813.1 | 2 - 3 | 176 | 70 | |
| GenBank mRNA | BC102369.1 | - | 2 - 3 | 178 | 70 | |
More Information
Set Membership: |
|

MRPL16
Genomic Map