Assay Details
| Gene Symbol: SEPTIN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034651.2 | NP_001029823.1 | 8 - 9 | 974 | 97 | |
| GenBank mRNA | BC102617.1 | - | 8 - 9 | 961 | 97 | |
| BC126719.1 | - | 7 - 8 | 1483 | 97 | ||
| BT029811.1 | - | 9 - 10 | 1444 | 97 | ||
More Information
Set Membership: |
|

SEPTIN4
Genomic Map