Assay Details
| Gene Symbol: PDZD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034664.1 | NP_001029836.1 | 3 - 4 | 124 | 61 | |
| GenBank mRNA | BC105272.1 | - | 3 - 4 | 124 | 61 | |
More Information
Set Membership: |
|

PDZD3
Genomic Map