Assay Details
| Gene Symbol: ASB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034669.1 | NP_001029841.1 | 6 - 7 | 1056 | 58 | |
| GenBank mRNA | BC104497.1 | - | 6 - 7 | 1056 | 58 | |
More Information
Set Membership: |
|

ASB2
Genomic Map