Assay Details
| Gene Symbol: APH1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034670.1 | NP_001029842.1 | 1 - 2 | 122 | 55 | |
| GenBank mRNA | BC103090.1 | - | 1 - 2 | 122 | 55 | |
More Information
Set Membership: |
|

APH1A
Genomic Map