Assay Details
| Gene Symbol: CALML4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034671.2 | NP_001029843.1 | 3 - 4 | 218 | 73 | |
| GenBank mRNA | BC102423.1 | - | 3 - 4 | 219 | 73 | |
More Information
Set Membership: |
|

CALML4
Genomic Map