Assay Details
| Gene Symbol: RPL14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034674.1 | NP_001029846.1 | 3 - 4 | 222 | 64 | |
| GenBank mRNA | BC102261.1 | - | 3 - 4 | 222 | 64 | |
| BC148013.1 | - | 3 - 4 | 222 | 64 | ||
More Information
Set Membership: |
|

RPL14
Genomic Map