Assay Details
| Gene Symbol: ILK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034693.1 | NP_001029865.1 | 2 - 3 | 167 | 90 | |
| GenBank mRNA | BC104603.1 | - | 2 - 3 | 167 | 90 | |
More Information
Set Membership: |
|

ILK
Genomic Map