Assay Details
| Gene Symbol: RAN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034705.2 | NP_001029877.1 | 6 - 7 | 657 | 61 | |
| GenBank mRNA | BC102559.1 | - | 6 - 7 | 658 | 61 | |
| BC151426.1 | - | 6 - 7 | 663 | 61 | ||
More Information
Set Membership: |
|

RAN
Genomic Map