Assay Details
| Gene Symbol: NR1H4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034708.2 | NP_001029880.1 | 8 - 9 | 1231 | 74 | |
| GenBank mRNA | BC102805.1 | - | 8 - 9 | 1232 | 74 | |
More Information
Set Membership: |
|

NR1H4
Genomic Map