Assay Details
| Gene Symbol: CCND3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034709.2 | NP_001029881.1 | 4 - 5 | 799 | 126 | |
| GenBank mRNA | BC105236.1 | - | 4 - 5 | 800 | 126 | |
More Information
Set Membership: |
|

CCND3
Genomic Map