Assay Details
| Gene Symbol: RAD21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034717.1 | NP_001029889.1 | 8 - 9 | 1123 | 73 | |
| GenBank mRNA | BC104612.1 | - | 8 - 9 | 1123 | 73 | |
More Information
Set Membership: |
|

RAD21
Genomic Map