Assay Details
| Gene Symbol: XBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034727.3 | NP_001029899.1 | 1 - 2 | 271 | 62 | |
| NM_001271737.1 | NP_001258666.1 | 1 - 2 | 271 | 62 | ||
| GenBank mRNA | BC102639.1 | - | 1 - 2 | 245 | 62 | |
| EH145309.1 | - | 1 - 2 | 272 | 62 | ||
| EH156163.1 | - | 1 - 2 | 65 | 62 | ||
More Information
Set Membership: |
|

XBP1
Genomic Map