Assay Details
Gene Symbol: THBS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034728.1 | NP_001029900.1 | 19 - 20 | 2678 | 57 | |
GenBank mRNA | BC104630.1 | - | 19 - 20 | 2678 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
