Assay Details
| Gene Symbol: AVPI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034760.2 | NP_001029932.1 | 2 - 3 | 757 | 102 | |
| GenBank mRNA | BC102744.1 | - | 2 - 3 | 758 | 102 | |
More Information
Set Membership: |
|

AVPI1
Genomic Map