Assay Details
| Gene Symbol: LGALS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034768.2 | NP_001029940.1 | 4 - 5 | 529 | 101 | |
| GenBank mRNA | BC102444.1 | - | 4 - 5 | 531 | 101 | |
More Information
Set Membership: |
|

LGALS4
Genomic Map