Assay Details
| Gene Symbol: HMGN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034772.1 | NP_001029944.1 | 3 - 4 | 176 | 117 | |
| GenBank mRNA | BC102449.1 | - | 3 - 4 | 176 | 117 | |
More Information
Set Membership: |
|

HMGN1
Genomic Map