Assay Details
| Gene Symbol: STMN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034790.2 | NP_001029962.1 | 4 - 5 | 473 | 70 | |
| GenBank mRNA | BC102453.1 | - | 4 - 5 | 474 | 70 | |
More Information
Set Membership: |
|

STMN1
Genomic Map