Assay Details
| Gene Symbol: GALM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034795.1 | NP_001029967.1 | 4 - 5 | 680 | 57 | |
| GenBank mRNA | BC102446.1 | - | 4 - 5 | 680 | 57 | |
| BT020690.1 | - | 4 - 5 | 697 | 57 | ||
More Information
Set Membership: |
|

GALM
Genomic Map