Assay Details
| Gene Symbol: TRAPPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034796.1 | NP_001029968.1 | 2 - 3 | 164 | 110 | |
| GenBank mRNA | BC102418.1 | - | 2 - 3 | 164 | 110 | |
More Information
Set Membership: |
|

TRAPPC2
Genomic Map