Assay Details
| Gene Symbol: RAB24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034802.1 | NP_001029974.1 | 7 - 8 | 1070 | 82 | |
| GenBank mRNA | BC102183.1 | - | 7 - 8 | 1070 | 82 | |
More Information
Set Membership: |
|

RAB24
Genomic Map